Gene Transcripts

Gene Symbol
UFD1L
Entrez Gene ID
7353
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Gene Aliases
UFD1
Chromosome Location
Chr. 22: 19449941 - 19479215 on Build GRCh38
UniGene ID
Hs.474213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035247.2 NP_001030324.2 9, 10 816
NM_005659.6 NP_005650.2 9, 10 816
XM_011530361.2 XP_011528663.1 9, 10 942
GenBank mRNA AF141201.1 - 8, 9 686
AJ239058.1 - 8, 9 686
AK128120.1 - 4, 5 2707
AK225877.1 - 9, 10 773
AK304734.1 - 9, 10 786
AK315998.1 - 9, 10 767
AY101594.1 - 9, 10 743
BC001049.2 - 9, 10 763 3507963
BC005087.1 - 9, 10 764 3830774
BX648483.1 - 9, 10 748
CR456607.1 - 9, 10 700
U64444.1 - 9, 10 872

View Full Product Details