Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077269.1 | NP_001070737.1 | 5 | 736 | |
NM_003387.4 | NP_003378.3 | 5 | 672 | ||
XM_011511780.2 | XP_011510082.1 | 5 | 858 | ||
GenBank mRNA | AF031588.1 | - | 5 | 680 | |
AK301271.1 | - | 5 | 677 | ||
AK304194.1 | - | 5 | 689 | ||
AL529597.3 | - | 5 | 704 | ||
BC002914.2 | - | 5 | 764 | 3954968 | |
BC110288.1 | - | 5 | 708 | 5184105 | |
BQ441937.1 | - | 3 | 473 | ||
BX640870.1 | - | 5 | 736 | ||
X86019.1 | - | 5 | 776 |