Gene Transcripts
- Gene Symbol
- ZNF79
- Entrez Gene ID
- 7633
- Gene Name
- zinc finger protein 79
- Gene Aliases
- pT7
- Chromosome Location
- Chr. 9: 127424374 - 127446289 on Build GRCh38
- UniGene ID
- Hs.522399
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286696.1 |
NP_001273625.1
|
2, 3 |
637 |
|
NM_001286697.1 |
NP_001273626.1
|
3, 4 |
639 |
|
NM_001322260.1 |
NP_001309189.1
|
3, 4 |
768 |
|
NM_007135.2 |
NP_009066.2
|
2, 3 |
530 |
|
XM_006717279.3 |
XP_006717342.1
|
3, 4 |
738 |
|
GenBank mRNA |
AK054606.1 |
- |
2, 3 |
522 |
|
AK302284.1 |
- |
2, 3 |
579 |
|
AK316233.1 |
- |
3, 4 |
575 |
|
BC062309.1 |
- |
2, 3 |
485 |
3944657 |
BP351823.1 |
- |
2, 3 |
492 |
|
View Full Product Details