Gene Transcripts

Gene Symbol
ST8SIA2
Entrez Gene ID
8128
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
HsT19690, SIAT8B, ST8SIA-II, STX
Chromosome Location
Chr. 15: 92393910 - 92468728 on Build GRCh38
UniGene ID
Hs.302341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006011.3 NP_006002.1 6 1198
XM_017022642.1 XP_016878131.1 6 1361
XM_017022643.1 XP_016878132.1 5 1134
GenBank mRNA AK313924.1 - 6 1146
BC069584.1 - 6 1067
BC096203.3 - 6 1067 40004645
BC096204.3 - 6 1067 40004646
BC096205.3 - 5 1004 40004648
L29556.1 - 3 584
U33551.1 - 6 1043
U82762.1 - 6 1068

View Full Product Details