Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006767.3 | NP_006758.2 | 19 | 2389 | |
GenBank mRNA | AK296347.1 | - | 18 | 2270 | |
AK313502.1 | - | 14 | 1791 | ||
AL833698.1 | - | 2 | 1584 | ||
BC026214.1 | - | 19 | 2291 | 4399716 | |
CT841521.1 | - | 19 | 2305 | ||
D38496.1 | - | 19 | 2284 |