Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003647.2 | NP_003638.1 | 6 | 1156 | |
XM_011525394.2 | XP_011523696.1 | 6 | 1134 | ||
XM_011525395.2 | XP_011523697.1 | 6 | 1141 | ||
XM_011525396.2 | XP_011523698.1 | 6 | 1143 | ||
XM_017025243.1 | XP_016880732.1 | 6 | 1523 | ||
XM_017025244.1 | XP_016880733.1 | 6 | 1137 | ||
XM_017025245.1 | XP_016880734.1 | 6 | 1140 | ||
GenBank mRNA | AB209365.1 | - | 5 | 806 | |
AK293039.1 | - | 6 | 1103 | ||
BC130629.1 | - | 5 | 1000 | 40146660 | |
BC130631.1 | - | 5 | 1000 | 40146662 | |
U49379.1 | - | 6 | 1063 |