Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003801.3 | NP_003792.1 | 5 | 649 | |
GenBank mRNA | AB002135.1 | - | 5 | 634 | |
AB006969.1 | - | 5 | 547 | ||
AK312759.1 | - | 5 | 661 | ||
AL157437.1 | - | 4 | 1146 | ||
BC003171.1 | - | 5 | 625 | 2906111 | |
BC004129.2 | - | 5 | 649 | 3504366 | |
BC006383.2 | - | 4 | 422 | 4099139 |