Gene Transcripts

Gene Symbol
TNFRSF14
Entrez Gene ID
8764
Gene Name
TNF receptor superfamily member 14
Gene Aliases
ATAR, CD270, HVEA, HVEM, LIGHTR, TR2
Chromosome Location
Chr. 1: 2555767 - 2565622 on Build GRCh38
UniGene ID
Hs.512898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003820.3 NP_003811.2 6, 7 1004
XM_006711019.2 XP_006711082.1 6, 7 676
XM_011542383.2 XP_011540685.1 3, 4 1047
XM_017002718.1 XP_016858207.1 3, 4 1127
XM_017002720.1 XP_016858209.1 6, 7 1722
XM_017002721.1 XP_016858210.1 7, 8 1132
GenBank mRNA AB208808.1 - 5 3380
AF153978.1 - 6, 7 949
AF373877.1 - 6, 7 704
AF373878.1 - 6, 7 704
AK124010.1 - 4, 5 1903
AY358879.1 - 6, 7 785
BC002794.2 - 6, 7 970 3614650
BC018882.2 - 6, 7 970
BC029848.1 - 4, 5 941 5178561
CD366981.1 - 2 716
CR456909.1 - 6, 7 704
U70321.1 - 6, 7 997
U81232.1 - 6, 7 968

View Full Product Details