Gene Transcripts
- Gene Symbol
- RGS11
- Entrez Gene ID
- 8786
- Gene Name
- regulator of G-protein signaling 11
- Gene Aliases
- RS11
- Chromosome Location
- Chr. 16: 268301 - 275943 on Build GRCh38
- UniGene ID
- Hs.65756
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286485.1 |
NP_001273414.1
|
5, 6 |
423 |
|
NM_001286486.1 |
NP_001273415.1
|
5, 6 |
360 |
|
NM_003834.2 |
NP_003825.1
|
6, 7 |
467 |
|
NM_183337.2 |
NP_899180.1
|
6, 7 |
456 |
|
XM_011522719.2 |
XP_011521021.1
|
7, 8 |
550 |
|
XM_011522720.2 |
XP_011521022.1
|
7, 8 |
490 |
|
XM_011522721.1 |
XP_011521023.1
|
7, 8 |
550 |
|
XM_011522722.1 |
XP_011521024.1
|
7, 8 |
550 |
|
XM_011522723.1 |
XP_011521025.1
|
6, 7 |
454 |
|
GenBank mRNA |
AB016929.1 |
- |
5, 6 |
391 |
|
AB209582.1 |
- |
1, 2 |
909 |
|
AF035153.1 |
- |
6, 7 |
427 |
|
AF035154.1 |
- |
6, 7 |
438 |
|
AK308972.1 |
- |
5, 6 |
677 |
|
View Full Product Details