Gene Transcripts

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
Chromosome Location
Chr. 16: 1333605 - 1349441 on Build GRCh38
UniGene ID
Hs.458427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199096.1 NP_001186025.1 14 1404
NM_001199097.1 NP_001186026.1 15 1454
NM_001199098.1 NP_001186027.1 15 1385
NM_001199099.1 NP_001186028.1 15 1370
NM_001286464.1 NP_001273393.1 15 1505
NM_003933.4 NP_003924.2 15 1562
XM_011522728.1 XP_011521030.1 15 2108
XM_011522729.1 XP_011521031.1 15 2108
XM_011522730.2 XP_011521032.1 15 2108
GenBank mRNA AB018277.2 - 15 1414
AK294576.1 - 4 325
AK294744.1 - 15 1330
AK295645.1 - 15 1344
AK299309.1 - 15 1414
AK299358.1 - 14 1404
AK315059.1 - 15 1562
AL834321.1 - 15 1456
BC104966.1 - 15 1425 8143969
BC112129.1 - 15 1437 8327597

View Full Product Details