Gene Transcripts

Gene Symbol
SLC25A14
Entrez Gene ID
9016
Gene Name
solute carrier family 25 member 14
Gene Aliases
BMCP1, UCP5
Chromosome Location
Chr. X: 130339888 - 130373361 on Build GRCh38
UniGene ID
Hs.194686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282195.1 NP_001269124.1 4 487
NM_001282196.1 NP_001269125.1 4 478
NM_001282197.1 NP_001269126.1 4 478
NM_001282198.1 NP_001269127.1 3 240
NR_104107.1 3 240
XM_011531402.2 XP_011529704.1 4 489
XM_011531403.1 XP_011529705.1 3 203
XM_017029936.1 XP_016885425.1 3 203
XM_017029937.1 XP_016885426.1 4 489
XM_017029938.1 XP_016885427.1 4 484
XM_017029939.1 XP_016885428.1 4 453
XM_017029941.1 XP_016885430.1 4 462
GenBank mRNA AF078544.1 - 3 483
AF155809.1 - 3 226
AF155810.1 - 3 217
AF155811.1 - 3 217
AK296155.1 - 3 170
AK297499.1 - 4 421
AK304649.1 - 3 220
AK309433.1 - 4 412
AL520510.3 - 3 190
AY358099.1 - 3 235
BC119666.2 - 3 342 40116117
BC119667.1 - 3 342 40116118
DB485834.2 - 3 240

View Full Product Details