Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003958.3 | NP_003949.1 | 5 | 1270 | |
NM_183078.2 | NP_898901.1 | 5 | 1270 | ||
NR_046399.1 | 5 | 1376 | |||
XM_017011462.1 | XP_016866951.1 | 5 | 1121 | ||
XM_017011463.1 | XP_016866952.1 | 5 | 1126 | ||
XM_017011464.1 | XP_016866953.1 | 5 | 1275 | ||
GenBank mRNA | AB014546.1 | - | 5 | 1189 | |
AF334675.1 | - | 5 | 1077 | ||
AK022075.1 | - | 5 | 1171 | ||
AK222765.1 | - | 5 | 1188 | ||
AK298319.1 | - | 5 | 1294 | ||
BC007517.2 | - | 5 | 1199 | 3028746 | |
BG029939.1 | - | 3 | 303 | ||
BT007446.1 | - | 5 | 1077 |