Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005443.4 | NP_005434.4 | 7 | 948 | |
XM_011532400.1 | XP_011530702.1 | 7 | 928 | ||
XM_011532401.1 | XP_011530703.1 | 7 | 1014 | ||
GenBank mRNA | AF016496.1 | - | 7 | 864 | |
AF033026.1 | - | 7 | 890 | ||
AF105227.1 | - | 7 | 918 | ||
AK292774.1 | - | 7 | 898 | ||
AK297981.1 | - | 7 | 771 | ||
BC011392.1 | - | 7 | 821 | 3869484 | |
BC032513.1 | - | 7 | 917 | ||
BC040940.1 | - | 7 | 876 | ||
BC050627.1 | - | 7 | 884 | 5556165 | |
CR457028.1 | - | 7 | 864 | ||
Y10387.1 | - | 7 | 900 |