Gene Transcripts

Gene Symbol
SLC24A1
Entrez Gene ID
9187
Gene Name
solute carrier family 24 member 1
Gene Aliases
CSNB1D, HsT17412, NCKX, NCKX1, RODX
Chromosome Location
Chr. 15: 65611405 - 65660995 on Build GRCh38
UniGene ID
Hs.173092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301031.1 NP_001287960.1 1 1788
NM_001301032.1 NP_001287961.1 1 1788
NM_001301033.1 NP_001287962.1 4 2286
NM_004727.2 NP_004718.1 2 1949
XM_005254778.3 XP_005254835.1 3 1923
XM_005254781.3 XP_005254838.1 3 1923
XM_006720768.3 XP_006720831.1 3 1923
XM_011522219.1 XP_011520521.1 3 2017
XM_011522220.2 XP_011520522.1 3 1923
XM_011522221.2 XP_011520523.1 3 1923
XM_011522222.2 XP_011520524.1 3 1923
XM_011522226.2 XP_011520528.1 3 1923
XM_017022724.1 XP_016878213.1 3 1923
XM_017022725.1 XP_016878214.1 3 1923
GenBank mRNA AB014602.1 - 3 3137
AF026132.1 - 1 1662
AF062921.1 - 1 1662
AF062922.1 - 2 1949
AK304006.1 - 4 2286
BC117263.1 - 1 1744 40125814
BC143375.1 - 1 1744 9051880

View Full Product Details