Gene Transcripts

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
Chromosome Location
Chr. 2: 96266146 - 96274179 on Build GRCh38
UniGene ID
Hs.12109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004804.2 NP_004795.1 6 931
GenBank mRNA AK056423.1 - 4 1297
AK299503.1 - 6 618
AK315568.1 - 6 840
BC001395.2 - 6 795 3050587
BC032812.1 - 6 825 5219740
CR456802.1 - 6 726
EF011618.1 - 6 825
U63810.1 - 6 882

View Full Product Details