Gene Transcripts

Gene Symbol
GSTO1
Entrez Gene ID
9446
Gene Name
glutathione S-transferase omega 1
Gene Aliases
GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
Chromosome Location
Chr. 10: 104254194 - 104267464 on Build GRCh38
UniGene ID
Hs.190028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191002.1 NP_001177931.1 4 626
NM_001191003.1 NP_001177932.1 5 778
NM_004832.2 NP_004823.1 5 725
GenBank mRNA AF212303.1 - 5 532
BC000127.1 - 5 535 3351359
BX537431.1 - 5 653
CA431506.1 - 2 289
CF130513.1 - 5 502
CN283947.1 - 4 429
EU794619.1 - 5 532
U90313.1 - 5 515

View Full Product Details