Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004297.3 | NP_004288.1 | 2 | 800 | |
XM_017015341.1 | XP_016870830.1 | 2 | 328 | ||
GenBank mRNA | AF105201.1 | - | 2 | 415 | |
AF493903.1 | - | 2 | 277 | ||
AK312460.1 | - | 2 | 800 | ||
BC027886.1 | - | 2 | 791 | 5221889 |