Gene Transcripts

Gene Symbol
TTC37
Entrez Gene ID
9652
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
KIAA0372, Ski3, THES
Chromosome Location
Chr. 5: 95463895 - 95555005 on Build GRCh38
UniGene ID
Hs.482868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014639.3 NP_055454.1 33 3787
GenBank mRNA AB002370.1 - 33 3787
BC056893.1 - 33 3760 6164578

View Full Product Details