Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
Chromosome Location
Chr. 6: 52497402 - 52577064 on Build GRCh38
UniGene ID
Hs.520182
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012288.3 NP_036420.1 1 251
GenBank mRNA AB621814.1 - 1 247
AK291751.1 - 1 137
BC028121.1 - 1 251 5169808
D31762.1 - 1 177
KU178488.1 - 1 102
KU178489.1 - 1 102

View Full Product Details