Gene Transcripts
- Gene Symbol
- MED16
- Entrez Gene ID
- 10025
- Gene Name
- mediator complex subunit 16
- Gene Aliases
- DRIP92, THRAP5, TRAP95
- Chromosome Location
- Chr. 19: 867961 - 893228 on Build GRCh38
- UniGene ID
- Hs.365207
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005481.2 |
NP_005472.2
|
6, 7 |
1144 |
|
XM_017026120.1 |
XP_016881609.1
|
6, 7 |
1139 |
|
XM_017026121.1 |
XP_016881610.1
|
5, 6 |
984 |
|
GenBank mRNA |
AF106934.1 |
- |
6, 7 |
1131 |
|
AF121228.1 |
- |
5, 6 |
993 |
|
BC004554.1 |
- |
6, 7 |
1125 |
4053743 |
BC007853.2 |
- |
2, 3 |
254 |
|
BC011841.1 |
- |
6, 7 |
1125 |
4053664 |
BC017282.2 |
- |
6, 7 |
1119 |
4876977 |
EU392449.1 |
- |
5, 6 |
1010 |
|
EU392451.1 |
- |
4, 5 |
572 |
|
View Full Product Details