Gene Transcripts

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
Chromosome Location
Chr. 6: 30151939 - 30163216 on Build GRCh38
UniGene ID
Hs.709483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006778.3 NP_006769.2 3, 4 839
NM_052828.2 NP_439893.2 3, 4 839
XM_011514221.1 XP_011512523.1 5, 6 1087
XM_011514222.2 XP_011512524.1 5, 6 1321
XM_011514223.2 XP_011512525.1 6, 7 1202
XM_011514225.1 XP_011512527.1 5, 6 1087
GenBank mRNA AF220122.1 - 3, 4 837
AF220123.1 - 3, 4 837
AL360163.1 - 1, 2 102
AL360164.1 - 1, 2 133
BC093926.1 - 3, 4 817 7939771
BC113419.1 - 3, 4 817 8322471

View Full Product Details