Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190979.2 | NP_001177908.1 | 2 | 305 | |
NM_001190980.2 | NP_001177909.1 | 2 | 305 | ||
NM_001320080.1 | NP_001307009.1 | 2 | 305 | ||
NM_005748.5 | NP_005739.2 | 2 | 305 | ||
NR_135139.1 | 2 | 305 | |||
NR_135140.1 | 2 | 305 | |||
XM_006719184.3 | XP_006719247.1 | 1 | 91 | ||
XM_011537728.2 | XP_011536030.1 | 2 | 230 | ||
XM_017018669.1 | XP_016874158.1 | 1 | 47 | ||
XM_017018670.1 | XP_016874159.1 | 2 | 243 | ||
GenBank mRNA | AI654632.1 | - | 4 | 299 | |
AK127531.1 | - | 2 | 185 | ||
AK291355.1 | - | 2 | 185 | ||
AK296191.1 | - | 2 | 185 | ||
BC037777.2 | - | 2 | 173 | 5276663 | |
CB963485.1 | - | 2 | 160 | ||
CB993159.1 | - | 2 | 184 | ||
DA011078.1 | - | 2 | 286 | ||
DA491895.1 | - | 2 | 289 | ||
HY111766.1 | - | 2 | 305 |