Gene Transcripts

Gene Symbol
UBE4B
Entrez Gene ID
10277
Gene Name
ubiquitination factor E4B
Gene Aliases
E4, HDNB1, UBOX3, UFD2, UFD2A
Chromosome Location
Chr. 1: 10032983 - 10181239 on Build GRCh38
UniGene ID
Hs.593974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105562.2 NP_001099032.1 13 2473
NM_006048.4 NP_006039.2 12 2086
XM_005263422.1 XP_005263479.1 14 2450
XM_011540488.2 XP_011538790.1 13 1749
XM_011540489.2 XP_011538791.1 14 2450
XM_011540490.1 XP_011538792.1 14 2450
XM_017000035.1 XP_016855524.1 11 1536
XM_017000036.1 XP_016855525.1 13 1750
GenBank mRNA AB014584.3 - 12 1977
AB028839.1 - 12 2094
AF043117.1 - 13 1870
AF331520.1 - 12 1445
AK023654.1 - 2 1200
AK292444.1 - 12 1568
AK304484.1 - 10 1073
BC093696.1 - 12 1476 7939541
BC143458.1 - 12 1476
BC143460.1 - 11 1361
JF289274.1 - 14 1938

View Full Product Details