Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006062.2 | NP_006053.2 | 13 | 1186 | |
XM_006711918.3 | XP_006711981.1 | 13 | 1273 | ||
XM_017003163.1 | XP_016858652.1 | 12 | 2906 | ||
GenBank mRNA | AK300779.1 | - | 13 | 1160 | |
AK301809.1 | - | 13 | 1179 | ||
BC073806.1 | - | 13 | 1162 | 6671194 | |
U50383.1 | - | 13 | 1139 |