Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016134.3 | NP_057218.1 | 2 | 526 | |
XM_017012953.1 | XP_016868442.1 | 2 | 526 | ||
XM_017012954.1 | XP_016868443.1 | 2 | 526 | ||
XM_017012955.1 | XP_016868444.1 | 2 | 526 | ||
GenBank mRNA | AF107833.1 | - | 1 | 87 | |
AF107834.1 | - | 1 | 369 | ||
AF119386.1 | - | 1 | 340 | ||
AK075132.1 | - | 2 | 444 | ||
AK315447.1 | - | 2 | 443 | ||
BC020689.1 | - | 2 | 482 | 4251802 |