Gene Transcripts

Gene Symbol
MAD2L2
Entrez Gene ID
10459
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Gene Aliases
MAD2B, POLZ2, REV7
Chromosome Location
Chr. 1: 11674480 - 11691788 on Build GRCh38
UniGene ID
Hs.19400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127325.1 NP_001120797.1 4 399
NM_006341.3 NP_006332.3 4 366
XM_011540507.1 XP_011538809.1 4 227
GenBank mRNA AF072933.1 - 4 210
AF080398.1 - 3 195
AF139365.1 - 4 289
AF157482.1 - 4 366
AK027327.1 - 4 279
AK094316.1 - 6 1108
BC015244.1 - 3 180 3916742
BI517654.1 - 4 399

View Full Product Details