Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006360.5 | NP_006351.2 | 3 | 365 | |
XM_011519840.2 | XP_011518142.1 | 3 | 389 | ||
GenBank mRNA | AF064603.1 | - | 3 | 295 | |
AK131064.1 | - | 3 | 319 | ||
AK222564.1 | - | 3 | 284 | ||
AK292139.1 | - | 3 | 307 | ||
AK312512.1 | - | 3 | 334 | ||
AY769947.1 | - | 3 | 244 | ||
BC019103.2 | - | 3 | 287 | 5021401 | |
BC051292.1 | - | 3 | 289 | 3607828 | |
CR450300.1 | - | 3 | 244 | ||
DB479224.1 | - | 3 | 365 | ||
DQ185042.1 | - | 3 | 235 |