Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006404.4 | NP_006395.2 | 3 | 628 | |
XM_011528496.1 | XP_011526798.1 | 3 | 471 | ||
GenBank mRNA | AK314887.1 | - | 3 | 594 | |
BC014451.1 | - | 3 | 492 | 4907433 | |
BM808358.1 | - | 3 | 646 | ||
CR456948.1 | - | 3 | 410 | ||
L35545.1 | - | 3 | 434 | ||
X89079.1 | - | 3 | 439 |