Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr. 1: 243255328 - 243500092 on Build GRCh38
UniGene ID
Hs.591530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006642.3 NP_006633.1 5 649
XM_005273013.4 XP_005273070.1 5 610
XM_005273018.1 XP_005273075.1 5 617
XM_005273021.4 XP_005273078.1 4 324
XM_005273022.3 XP_005273079.1 5 653
XM_005273023.4 XP_005273080.1 5 1326
XM_006711727.3 XP_006711790.1 5 535
XM_006711728.3 XP_006711791.1 5 535
XM_011544021.2 XP_011542323.1 5 535
XM_011544022.2 XP_011542324.1 5 610
XM_011544023.2 XP_011542325.1 5 535
XM_011544024.2 XP_011542326.1 5 535
XM_011544026.2 XP_011542328.1 5 535
XM_011544028.2 XP_011542330.1 5 535
XM_011544029.2 XP_011542331.1 5 535
XM_017000103.1 XP_016855592.1 5 535
XM_017000104.1 XP_016855593.1 5 610
XM_017000105.1 XP_016855594.1 5 636
XM_017000106.1 XP_016855595.1 5 617
XM_017000107.1 XP_016855596.1 4 324
XM_017000108.1 XP_016855597.1 5 610
XM_017000109.1 XP_016855598.1 5 535
GenBank mRNA AF161348.1 - 5 626
AF250731.1 - 5 480
AK297105.1 - 2 197
AK299212.1 - 5 598
AK299249.1 - 2 241
BC032454.1 - 5 592 5174799

View Full Product Details