Gene Transcripts

Gene Symbol
PRDM5
Entrez Gene ID
11107
Gene Name
PR domain 5
Gene Aliases
BCS2, PFM2
Chromosome Location
Chr. 4: 120684907 - 120922866 on Build GRCh38
UniGene ID
Hs.666782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300823.1 NP_001287752.1 4 620
NM_001300824.1 NP_001287753.1 4 620
NM_018699.3 NP_061169.2 4 620
XM_005262708.3 XP_005262765.1 4 603
XM_011531562.2 XP_011529864.1 4 603
XM_011531563.2 XP_011529865.1 4 603
XM_011531564.2 XP_011529866.1 4 603
XM_011531565.2 XP_011529867.1 4 603
XM_011531566.2 XP_011529868.1 4 603
XM_011531567.2 XP_011529869.1 4 603
XM_011531568.2 XP_011529870.1 4 603
XM_011531569.2 XP_011529871.1 4 603
XM_011531570.2 XP_011529872.1 4 435
XM_017007668.1 XP_016863157.1 4 1096
XM_017007669.1 XP_016863158.1 4 603
XM_017007670.1 XP_016863159.1 4 603
GenBank mRNA AF272897.1 - 4 458
AK310958.1 - 4 449
AK314293.1 - 4 436
AW341546.1 - 4 383
BC121037.1 - 4 407 40117856
BC121038.1 - 4 407 40117857

View Full Product Details