Gene Transcripts

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
Chromosome Location
Chr. 6: 105096822 - 105137174 on Build GRCh38
UniGene ID
Hs.221660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199563.1 NP_001186492.1 4 598
NM_007073.4 NP_009004.2 4 468
NM_147147.3 NP_671488.1 4 538
XM_011535398.2 XP_011533700.1 4 473
GenBank mRNA AK289979.1 - 4 582
BC034425.1 - 4 538 4821891
BC040502.1 - 4 598 5295907

View Full Product Details