Gene Transcripts
- Gene Symbol
- WDHD1
- Entrez Gene ID
- 11169
- Gene Name
- WD repeat and HMG-box DNA binding protein 1
- Gene Aliases
- AND-1, AND1, CHTF4, CTF4
- Chromosome Location
- Chr. 14: 54938938 - 55027105 on Build GRCh38
- UniGene ID
- Hs.385998
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008396.2 |
NP_001008397.1
|
12, 13 |
1533 |
|
NM_007086.3 |
NP_009017.1
|
13, 14 |
1626 |
|
XM_006720012.2 |
XP_006720075.1
|
13, 14 |
1624 |
|
XM_011536373.2 |
XP_011534675.1
|
12, 13 |
1425 |
|
XM_017020940.1 |
XP_016876429.1
|
12, 13 |
1537 |
|
XM_017020941.1 |
XP_016876430.1
|
10, 11 |
1235 |
|
GenBank mRNA |
AJ006266.1 |
- |
12, 13 |
1575 |
|
AK001538.1 |
- |
1, 2 |
180 |
|
AK001585.1 |
- |
3, 4 |
549 |
|
AK293005.1 |
- |
13, 14 |
1593 |
|
BC000622.2 |
- |
13, 14 |
1591 |
|
BC005040.1 |
- |
13, 14 |
1591 |
|
BC043349.1 |
- |
13, 14 |
1614 |
|
BC063041.1 |
- |
13, 14 |
1602 |
6514641 |
View Full Product Details