Gene Transcripts

Gene Symbol
NISCH
Entrez Gene ID
11188
Gene Name
nischarin
Gene Aliases
I-1, IR1, IRAS, hIRAS
Chromosome Location
Chr. 3: 52455508 - 52493072 on Build GRCh38
UniGene ID
Hs.435290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276293.1 NP_001263222.1 2 259
NM_001276294.1 NP_001263223.1 2 259
NM_007184.3 NP_009115.2 2 259
XM_005264839.2 XP_005264896.1 1 32
GenBank mRNA AB023192.1 - 3 197
AF082516.1 - 2 151
AK027428.1 - 2 137
AK291398.1 - 2 163
AK297657.1 - 2 163
AK315001.1 - 2 163
AL703940.1 - 2 133
BC038102.2 - 2 163 5268247
BC054494.1 - 2 157 6195532
BC056900.1 - 2 129 6143316
DB496030.1 - 2 259

View Full Product Details