Gene Transcripts
- Gene Symbol
- MTF2
- Entrez Gene ID
- 22823
- Gene Name
- metal response element binding transcription factor 2
- Gene Aliases
- M96, PCL2, TDRD19A, dJ976O13.2
- Chromosome Location
- Chr. 1: 93079219 - 93139081 on Build GRCh38
- UniGene ID
- Hs.31016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164391.1 |
NP_001157863.1
|
11, 12 |
1208 |
|
NM_001164393.1 |
NP_001157865.1
|
8, 9 |
1007 |
|
NM_007358.3 |
NP_031384.1
|
10, 11 |
1288 |
|
XM_011541016.1 |
XP_011539318.1
|
10, 11 |
1244 |
|
XM_011541019.1 |
XP_011539321.1
|
10, 11 |
1036 |
|
XM_017000676.1 |
XP_016856165.1
|
10, 11 |
1143 |
|
XM_017000679.1 |
XP_016856168.1
|
9, 10 |
1120 |
|
GenBank mRNA |
AF072814.1 |
- |
9, 10 |
986 |
|
AF086908.1 |
- |
9, 10 |
1089 |
|
AJ010014.1 |
- |
10, 11 |
1239 |
|
AK290318.1 |
- |
10, 11 |
1240 |
|
AK302776.1 |
- |
11, 12 |
1147 |
|
AK302900.1 |
- |
8, 9 |
959 |
|
BC001897.1 |
- |
10, 11 |
1207 |
|
View Full Product Details