Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
Chromosome Location
Chr. 15: 101175723 - 101251932 on Build GRCh38
UniGene ID
Hs.110488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014918.4 NP_055733.2 3 1435
XM_006720435.3 XP_006720498.1 3 717
XM_011521364.2 XP_011519666.1 4 1058
XM_017022011.1 XP_016877500.1 2 491
GenBank mRNA AB023207.1 - 3 1453
AB071402.1 - 3 973
AK296943.1 - 2 212
AY358529.1 - 3 1447
BC046247.1 - 3 1442 5751618

View Full Product Details