Gene Transcripts

Gene Symbol
ARHGEF15
Entrez Gene ID
22899
Gene Name
Rho guanine nucleotide exchange factor 15
Gene Aliases
ARGEF15, E5, Ephexin5, Vsm-RhoGEF
Chromosome Location
Chr. 17: 8310238 - 8322516 on Build GRCh38
UniGene ID
Hs.443109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025014.1 NP_079290.1 16 2573
NM_173728.3 NP_776089.2 16 2637
XM_011523734.1 XP_011522036.1 16 2682
XM_011523735.1 XP_011522037.1 16 2618
XM_011523736.1 XP_011522038.1 16 2633
XM_011523737.1 XP_011522039.1 13 1674
GenBank mRNA BQ877184.1 - 3 311
KU178557.1 - 15 2482
KU178558.1 - 14 2329

View Full Product Details