Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018158.2 | NP_060628.2 | 10 | 2194 | |
GenBank mRNA | AK001486.1 | - | 10 | 2194 | |
AK300958.1 | - | 12 | 1704 | ||
AY028435.1 | - | 10 | 2170 | ||
AY117688.1 | - | 10 | 1583 | ||
BC098302.1 | - | 10 | 2189 | 40026925 | |
BC098358.1 | - | 10 | 2189 | 40026922 | |
BC099711.1 | - | 10 | 2189 | 40026923 | |
BC099739.1 | - | 10 | 2187 | 40026926 |