Gene Transcripts

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
Chromosome Location
Chr. 10: 274190 - 689710 on Build GRCh38
UniGene ID
Hs.432397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014974.2 NP_055789.1 20 2396
XM_005252426.3 XP_005252483.1 21 2555
XM_005252427.3 XP_005252484.1 20 2436
XM_005252428.4 XP_005252485.1 21 2606
XM_005252430.2 XP_005252487.2 4 646
XM_011519428.2 XP_011517730.1 21 2537
XM_011519429.2 XP_011517731.1 21 2535
XM_011519430.2 XP_011517732.1 20 2412
XM_011519431.2 XP_011517733.1 20 2412
XM_011519432.1 XP_011517734.1 20 2837
XM_011519433.2 XP_011517735.1 4 1645
XM_017015977.1 XP_016871466.1 4 657
GenBank mRNA AB023151.2 - 20 2394
AK126063.1 - 22 2649
AK298353.1 - 4 493
AK309715.1 - 4 408
AK310018.1 - 3 649
BC063313.1 - 20 2396 30349267

View Full Product Details