Gene Transcripts

Gene Symbol
SEL1L3
Entrez Gene ID
23231
Gene Name
SEL1L family member 3
Gene Aliases
Sel-1L3
Chromosome Location
Chr. 4: 25697790 - 25863595 on Build GRCh38
UniGene ID
Hs.479384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297592.1 NP_001284521.1 15 2290
NM_001297594.1 NP_001284523.1 15 2312
NM_015187.4 NP_056002.2 15 2507
XM_011513819.2 XP_011512121.2 15 2501
XM_017007923.1 XP_016863412.1 14 2145
GenBank mRNA AB018289.1 - 14 2005
AK025282.1 - 3 173
AK289687.1 - 15 2255
AK300219.1 - 6 605
BC009945.2 - 8 1022
BC029422.1 - 5 432
BC041046.1 - 6 659
BC060867.1 - 15 2300 30344157
CR749817.1 - 15 2290

View Full Product Details