Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318326.1 | NP_001305255.1 | 4 | 1720 | |
NM_001318327.1 | NP_001305256.1 | 4 | 471 | ||
NM_015221.3 | NP_056036.1 | 7 | 2717 | ||
XM_006717735.2 | XP_006717798.1 | 7 | 2712 | ||
XM_011539559.2 | XP_011537861.1 | 8 | 2788 | ||
GenBank mRNA | AB023227.1 | - | 4 | 1807 | |
AK128576.1 | - | 4 | 1719 | ||
AL833283.1 | - | 7 | 2683 | ||
AY196211.1 | - | 6 | 2591 | ||
BC041628.1 | - | 4 | 460 | 4329464 |