Gene Transcripts

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
Chromosome Location
Chr. 2: 62673851 - 63046487 on Build GRCh38
UniGene ID
Hs.271667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015252.3 NP_056067.2 8 1188
XM_005264225.2 XP_005264282.1 8 1086
XM_005264226.2 XP_005264283.1 8 1443
XM_005264228.2 XP_005264285.1 8 1147
XM_006711971.2 XP_006712034.1 8 1147
XM_011532713.2 XP_011531015.1 8 1229
XM_011532714.2 XP_011531016.1 8 1078
XM_011532715.2 XP_011531017.1 8 1054
XM_011532716.2 XP_011531018.1 8 1147
XM_011532717.1 XP_011531019.1 8 1147
XM_011532718.2 XP_011531020.1 7 808
XM_017003643.1 XP_016859132.1 8 1044
XM_017003644.1 XP_016859133.1 8 1021
XM_017003645.1 XP_016859134.1 8 1086
XM_017003646.1 XP_016859135.1 8 1149
XM_017003650.1 XP_016859139.1 8 1086
XM_017003654.1 XP_016859143.1 8 1147
GenBank mRNA AL833968.1 - 8 1054
AY331186.1 - 8 1083
BP361773.1 - 3 345

View Full Product Details