Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017524.2 | NP_001017524.1 | 2 | 270 | |
NM_012239.5 | NP_036371.1 | 2 | 416 | ||
XM_005252835.1 | XP_005252892.1 | 2 | 416 | ||
XM_011519956.1 | XP_011518258.1 | 2 | 1208 | ||
XM_011519957.1 | XP_011518259.1 | 2 | 1037 | ||
XM_017017428.1 | XP_016872917.1 | 2 | 262 | ||
XM_017017429.1 | XP_016872918.1 | 2 | 436 | ||
XM_017017430.1 | XP_016872919.1 | 2 | 430 | ||
XM_017017431.1 | XP_016872920.1 | 2 | 161 | ||
GenBank mRNA | AF083108.2 | - | 2 | 484 | |
AK299009.1 | - | 2 | 406 | ||
AK299438.1 | - | 2 | 248 | ||
AK301979.1 | - | 2 | 404 | ||
AK308681.1 | - | 2 | 417 | ||
AK316175.1 | - | 2 | 320 | ||
AL535769.3 | - | 2 | 251 | ||
BC001042.2 | - | 2 | 399 | 2823471 | |
BI755839.1 | - | 2 | 216 |