Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014321.3 | NP_055136.1 | 3 | 340 | |
NR_037620.1 | 3 | 399 | |||
XM_011522978.2 | XP_011521280.1 | 3 | 330 | ||
GenBank mRNA | AF139658.1 | - | 3 | 328 | |
AK021946.1 | - | 3 | 301 | ||
AK024019.1 | - | 3 | 334 | ||
AK024077.1 | - | 3 | 327 | ||
AK310633.1 | - | 3 | 363 | ||
BC039032.1 | - | 3 | 320 | 5577396 | |
BC063565.1 | - | 3 | 309 | 3460593 | |
BG822640.1 | - | 3 | 311 |