Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282738.1 | NP_001269667.1 | 8 | 1332 | |
NM_001282739.1 | NP_001269668.1 | 7 | 1652 | ||
NM_001282740.1 | NP_001269669.1 | 6 | 835 | ||
NM_001282741.1 | NP_001269670.1 | 6 | 892 | ||
NM_001282742.1 | NP_001269671.1 | 5 | 563 | ||
NM_030758.3 | NP_110385.1 | 7 | 1655 | ||
XM_006724207.2 | XP_006724270.1 | 6 | 838 | ||
XM_006724208.3 | XP_006724271.1 | 5 | 623 | ||
XM_011530057.1 | XP_011528359.1 | 7 | 1551 | ||
XM_011530058.1 | XP_011528360.1 | 7 | 1548 | ||
XM_011530059.2 | XP_011528361.1 | 7 | 1581 | ||
XM_011530060.1 | XP_011528362.1 | 6 | 892 | ||
XM_011530061.1 | XP_011528363.1 | 5 | 536 | ||
XM_011530062.1 | XP_011528364.1 | 7 | 937 | ||
XM_017028709.1 | XP_016884198.1 | 7 | 1578 | ||
XM_017028710.1 | XP_016884199.1 | 7 | 1044 | ||
XM_017028712.1 | XP_016884201.1 | 6 | 839 | ||
XM_017028713.1 | XP_016884202.1 | 6 | 1165 | ||
GenBank mRNA | AB051451.2 | - | 7 | 1563 | |
AF288741.1 | - | 7 | 1549 | ||
AF323731.1 | - | 7 | 1655 | ||
AK093196.1 | - | 6 | 835 | ||
AK131374.1 | - | 7 | 1182 | ||
AK296354.1 | - | 6 | 892 | ||
AK296525.1 | - | 8 | 1334 | ||
AK300323.1 | - | 5 | 563 | ||
BC004340.1 | - | 4 | 450 | ||
BC118914.2 | - | 7 | 1569 | 40037933 |