Gene Transcripts

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
Chromosome Location
Chr. 11: 64035970 - 64119183 on Build GRCh38
UniGene ID
Hs.584876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013280.4 NP_037412.2 2 1843
XM_005273861.1 XP_005273918.1 3 2729
XM_011544883.1 XP_011543185.1 2 1683
GenBank mRNA AF007139.1 - 1 218
AF169675.1 - 1 1502
AK289931.1 - 2 2160
AY358308.1 - 2 1964
BC018370.1 - 2 1843 4214494

View Full Product Details