Gene Transcripts

Gene Symbol
PGLS
Entrez Gene ID
25796
Gene Name
6-phosphogluconolactonase
Gene Aliases
6PGL, HEL-S-304
Chromosome Location
Chr. 19: 17511623 - 17521291 on Build GRCh38
UniGene ID
Hs.466165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012088.2 NP_036220.1 4 564
XM_011527897.2 XP_011526199.1 4 1164
GenBank mRNA AF091091.1 - 3 664
AJ243972.1 - 4 531
BC014006.2 - 4 538 4053022
BG764228.1 - 4 562
GQ472229.1 - 4 538

View Full Product Details