Gene Transcripts

Gene Symbol
PGLS
Entrez Gene ID
25796
Gene Name
6-phosphogluconolactonase
Gene Aliases
6PGL, HEL-S-304
Chromosome Location
Chr. 19: 17511623 - 17521291 on Build GRCh38
UniGene ID
Hs.466165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012088.2 NP_036220.1 4 618
XM_011527897.2 XP_011526199.1 4 1218
GenBank mRNA AF091091.1 - 3 718
AJ243972.1 - 4 585

View Full Product Details