Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015397.3 | NP_056212.1 | 9 | 1630 | |
XM_011517843.1 | XP_011516145.1 | 9 | 1457 | ||
GenBank mRNA | AB067479.1 | - | 9 | 1550 | |
AF131771.1 | - | 4 | 490 | ||
AK000354.1 | - | 9 | 1408 | ||
AK292935.1 | - | 9 | 1436 | ||
AL117646.1 | - | 6 | 764 | ||
BC008893.2 | - | 8 | 1193 | ||
BC063823.1 | - | 9 | 1548 | 6184796 | |
DQ518909.1 | - | 9 | 1550 |