Gene Transcripts

Gene Symbol
KIF1BP
Entrez Gene ID
26128
Gene Name
KIF1 binding protein
Gene Aliases
KBP, KIAA1279, TTC20
Chromosome Location
Chr. 10: 68988721 - 69016983 on Build GRCh38
UniGene ID
Hs.279580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015634.3 NP_056449.1 2 596
GenBank mRNA AB033105.1 - 2 519
AK291343.1 - 2 501
BC006427.1 - 2 519
BC012180.1 - 2 493 4550085

View Full Product Details