Gene Transcripts
- Gene Symbol
- RGS22
- Entrez Gene ID
- 26166
- Gene Name
- regulator of G-protein signaling 22
- Gene Aliases
- CT145, PRTD-NY2
- Chromosome Location
- Chr. 8: 99960936 - 100106116 on Build GRCh38
- UniGene ID
- Hs.120021
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286692.1 |
NP_001273621.1
|
5, 6 |
623 |
|
NM_015668.4 |
NP_056483.3
|
5, 6 |
623 |
|
XM_005250856.2 |
XP_005250913.1
|
5, 6 |
469 |
|
XM_005250857.2 |
XP_005250914.1
|
5, 6 |
469 |
|
XM_005250858.2 |
XP_005250915.1
|
5, 6 |
469 |
|
XM_005250860.2 |
XP_005250917.1
|
5, 6 |
469 |
|
XM_006716540.2 |
XP_006716603.1
|
5, 6 |
737 |
|
XM_011516957.1 |
XP_011515259.1
|
6, 7 |
918 |
|
XM_011516958.1 |
XP_011515260.1
|
5, 6 |
816 |
|
XM_011516959.2 |
XP_011515261.1
|
5, 6 |
469 |
|
XM_017013309.1 |
XP_016868798.1
|
5, 6 |
469 |
|
XM_017013310.1 |
XP_016868799.1
|
5, 6 |
469 |
|
GenBank mRNA |
AK292559.1 |
- |
5, 6 |
469 |
|
AK309194.1 |
- |
2, 3 |
230 |
|
AY009106.1 |
- |
5, 6 |
452 |
|
BC036665.1 |
- |
5, 6 |
474 |
5268331 |
BC047060.1 |
- |
5, 6 |
507 |
|
BC092411.1 |
- |
5, 6 |
620 |
30386231 |
View Full Product Details